Skip to Content
Merck
All Photos(1)

Key Documents

870510C

Avanti

16:0 CDP DG

Avanti Research - A Croda Brand 870510C

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-(cytidine diphosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$737.00

$737.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$737.00

About This Item

Empirical Formula (Hill Notation):
C44H87N5O15P2
CAS Number:
Molecular Weight:
988.13
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$737.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 1 mL (870510C-10mg)
pkg of 1 × 1 mL (870510C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870510C

concentration

10 mg/mL (870510C-10mg)
5 mg/mL (870510C-5mg)

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(OP(OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C(N=C(N)C=C2)=O)O1)([O-])=O)([O-])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C44H81N3O15P2.2H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-39(48)57-33-36(60-40(49)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)34-58-63(53,54)62-64(55,56)59-35-37-41(50)42(51)43(61-37)47-32-31-38(45)46-44(47)52;;/h31-32,36-37,41-43,50-51H,3-30,33-35H2,1-2H3,(H,53,54)(H,55,56)(H2,45,46,52);2*1H3/t36-,37-,41+,42?,43-;;/m1../s1

InChI key

IAUUMXARBJMHDT-LWJUINKDSA-N

Application

Cytidine diphosphate-diacylglycerol (CDP-DG) is suitable for liposome preparation.[1]

Biochem/physiol Actions

Cytidine diphosphate-diacylglycerol (CDP-DG) acts as a precursor for synthesis of phosphatidylglycerol (PG) and phosphatidylinositol (PI) in alveolar type II cells.[2]

Packaging

5 mL Clear Glass Sealed Ampule (870510C-10mg)
5 mL Clear Glass Sealed Ampule (870510C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changes in phospholipids in bronchoalveolar lavage fluid of patients with interstitial lung diseases
Honda Y, et al.
Lung, 166(1), 293-301 (1988)
A steric gating mechanism dictates the substrate specificity of a Rab-GEF
Thomas L, et al.
Developmental Cell, 48(1), 100-114 (2019)
Julia Alvarez-Malmagro et al.
Bioelectrochemistry (Amsterdam, Netherlands), 132, 107416-107416 (2020-01-26)
A hybrid bilayer lipid membrane (hBLM), constructed with a 1-hexadecanethiol self-assembled interior leaflet and a 1,2-dipalmitoyl-sn-glycero-3-cytidine nucleolipid exterior leaflet, was deposited at the surface of a gold (111) electrode. This system was used to investigate the molecular recognition reaction between
Ru-Yin Huang et al.
Current microbiology, 77(5), 710-715 (2020-01-04)
Phosphatidylserine synthase (Pss) is involved in the metabolic pathway in phospholipid synthesis in different organisms. In this study, Pss expression in Vibrio parahaemolyticus was verified through liquid chromatography tandem-mass spectrometry. To analyze the characteristics of Pss, the recombinant Pss was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service