Skip to Content
Merck
All Photos(1)

Key Documents

870510P

Avanti

16:0 CDP DG

Avanti Research - A Croda Brand 870510P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-(cytidine diphosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$484.00

$484.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$484.00

About This Item

Empirical Formula (Hill Notation):
C44H87N5O15P2
CAS Number:
Molecular Weight:
988.13
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$484.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870510P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870510P

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(OP(OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C(N=C(N)C=C2)=O)O1)([O-])=O)([O-])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C44H81N3O15P2.2H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-39(48)57-33-36(60-40(49)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)34-58-63(53,54)62-64(55,56)59-35-37-41(50)42(51)43(61-37)47-32-31-38(45)46-44(47)52;;/h31-32,36-37,41-43,50-51H,3-30,33-35H2,1-2H3,(H,53,54)(H,55,56)(H2,45,46,52);2*1H3/t36-,37-,41+,42?,43-;;/m1../s1

InChI key

IAUUMXARBJMHDT-LWJUINKDSA-N

Application

16:0 CDP DG (1,2-dipalmitoyl-sn-glycero-3-(cytidine diphosphate)) may be used in liposome preparation.[1]

Biochem/physiol Actions

16:0 CDP DG (1,2-dipalmitoyl-sn-glycero-3-(cytidine diphosphate)) acts as a substrate for phosphatidylserine synthase (Pss) to produce cytidine 5′?monophosphate.[2] CDP-diacylglycerol is a derivate of diacylglycerol. It serves as a precursor for the synthesis of essential glycerophospholipids such as the phosphatidylinositides.[3] Altered CDP-diacylglycerol signaling might lead to depression.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (870510P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly R Tyeryar et al.
BMC neuroscience, 9, 12-12 (2008-01-26)
Major depression is a serious mood disorder affecting millions of adults and children worldwide. While the etiopathology of depression remains obscure, antidepressant medications increase synaptic levels of monoamine neurotransmitters in brain regions associated with the disease. Monoamine transmitters activate multiple
Julia Alvarez-Malmagro et al.
Bioelectrochemistry (Amsterdam, Netherlands), 132, 107416-107416 (2020-01-26)
A hybrid bilayer lipid membrane (hBLM), constructed with a 1-hexadecanethiol self-assembled interior leaflet and a 1,2-dipalmitoyl-sn-glycero-3-cytidine nucleolipid exterior leaflet, was deposited at the surface of a gold (111) electrode. This system was used to investigate the molecular recognition reaction between
Laura L Thomas et al.
Developmental cell, 48(1), 100-114 (2018-12-12)
Correct localization of Rab GTPases in cells is critical for proper function in membrane trafficking, yet the mechanisms that target Rabs to specific subcellular compartments remain controversial. Guanine nucleotide exchange factors (GEFs) activate and consequently stabilize Rab substrates on membranes
Ru-Yin Huang et al.
Current microbiology, 77(5), 710-715 (2020-01-04)
Phosphatidylserine synthase (Pss) is involved in the metabolic pathway in phospholipid synthesis in different organisms. In this study, Pss expression in Vibrio parahaemolyticus was verified through liquid chromatography tandem-mass spectrometry. To analyze the characteristics of Pss, the recombinant Pss was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service