Skip to Content
Merck
All Photos(1)

Documents

870520P

Avanti

18:1 CDP DG

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleoyl-sn-glycero-3-(cytidine diphosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H91N5O15P2
CAS Number:
Molecular Weight:
1040.21
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 25 mg (870520P-25mg)
pkg of 1 × 5 mg (870520P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP(OP(OC[C@@H]1[C@@H](O)[C@@H](O)[C@H](N2C(N=C(N)C=C2)=O)O1)([O-])=O)([O-])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[NH4+].[NH4+]

InChI

1S/C48H85N3O15P2.2H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-43(52)61-37-40(64-44(53)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)38-62-67(57,58)66-68(59,60)63-39-41-45(54)46(55)47(65-41)51-36-35-42(49)50-48(51)56;;/h17-20,35-36,40-41,45-47,54-55H,3-16,21-34,37-39H2,1-2H3,(H,57,58)(H,59,60)(H2,49,50,56);2*1H3/b19-17-,20-18-;;/t40-,41-,45+,46?,47-;;/m1../s1

InChI key

NECNTGAHQCHVGS-BQGZBBTBSA-N

Application

18:1 CDP DG (1,2-dioleoyl-sn-glycero-3-(cytidine diphosphate)) may be used in preparation of liposomes.

Biochem/physiol Actions

18:1 CDP DG (1,2-dioleoyl-sn-glycero-3-(cytidine diphosphate)) is a diacylglycerol derivate. It acts as a substrate for phosphatidylserine synthase (Pss), an enzyme involved in the synthesis of phospholipids in various organisms. CDP-diacylglycerol serves as a precursor for the biosynthesis of phosphatidylinositides. Dysregulated CDP-diacylglycerol signaling pathway leads to depression.

Packaging

5 mL Clear Glass Sealed Ampule (870520P-25mg)
5 mL Clear Glass Sealed Ampule (870520P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly R Tyeryar et al.
BMC neuroscience, 9, 12-12 (2008-01-26)
Major depression is a serious mood disorder affecting millions of adults and children worldwide. While the etiopathology of depression remains obscure, antidepressant medications increase synaptic levels of monoamine neurotransmitters in brain regions associated with the disease. Monoamine transmitters activate multiple
Andrew G Reidenbach et al.
Cell chemical biology, 25(2), 154-165 (2017-12-05)
Human COQ8A (ADCK3) and Saccharomyces cerevisiae Coq8p (collectively COQ8) are UbiB family proteins essential for mitochondrial coenzyme Q (CoQ) biosynthesis. However, the biochemical activity of COQ8 and its direct role in CoQ production remain unclear, in part due to lack
Ru-Yin Huang et al.
Current microbiology, 77(5), 710-715 (2020-01-04)
Phosphatidylserine synthase (Pss) is involved in the metabolic pathway in phospholipid synthesis in different organisms. In this study, Pss expression in Vibrio parahaemolyticus was verified through liquid chromatography tandem-mass spectrometry. To analyze the characteristics of Pss, the recombinant Pss was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service