Skip to Content
Merck
All Photos(1)

Key Documents

860582P

Avanti

06:0 SM (d18:1/6:0)

Avanti Research - A Croda Brand 860582P, powder

Synonym(s):

N-hexanoyl-D-erythro-sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$914.00

$914.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$914.00

About This Item

Empirical Formula (Hill Notation):
C29H59N2O6P
CAS Number:
Molecular Weight:
562.76
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$914.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860582P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860582P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C29H59N2O6P/c1-6-8-10-11-12-13-14-15-16-17-18-19-21-22-28(32)27(30-29(33)23-20-9-7-2)26-37-38(34,35)36-25-24-31(3,4)5/h21-22,27-28,32H,6-20,23-26H2,1-5H3,(H-,30,33,34,35)/b22-21+/t27-,28+/m0/s1

InChI key

AHZZHULAOVWYNO-RRKDMDGFSA-N

Application

06:0 SM (d18:1/6:0) may be used:
  • to study its effect on C-X-C motif chemokine ligand 12 (CXCL12)-induced cell migration using a transwell migration assay[1]
  • in liposomes to measure ostreolysin A (OlyA) binding in order to infer the effects of phosphatidylcholines (PCs) on sphingomyelin (SM)/cholesterol interactions[2]
  • as a component of lipid substrate mixture for substrate specificity studies of neutral sphingomyelinase fromTrypanosoma brucei[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860582P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily A Dickie et al.
Parasitology, 146(5), 604-616 (2018-11-06)
The kinetoplastid parasite Trypanosoma brucei causes African trypanosomiasis in both humans and animals. Infections place a significant health and economic burden on developing nations in sub-Saharan Africa, but few effective anti-parasitic treatments are currently available. Hence, there is an urgent
Satoshi Asano et al.
Molecular and cellular biology, 32(16), 3242-3252 (2012-06-13)
Sphingomyelin synthase (SMS) catalyzes the formation of sphingomyelin, a major component of the plasma membrane and lipid rafts. To investigate the role of SMS in cell signaling and migration induced by binding of the chemokine CXCL12 to CXCR4, we used
Shreya Endapally et al.
Cell, 176(5), 1040-1053 (2019-02-05)
Sphingomyelin and cholesterol are essential lipids that are enriched in plasma membranes of animal cells, where they interact to regulate membrane properties and many intracellular signaling processes. Despite intense study, the interaction between these lipids in membranes is not well
Delphine Milhas et al.
FEBS letters, 584(9), 1887-1894 (2009-10-28)
The plasma membrane (PM) is a major resource for production of bioactive lipids and contains a large proportion of the cellular sphingomyelin (SM) content. Consequently, the regulation of SM levels at the PM by enzymes such as sphingomyelinase (SMase) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service