Skip to Content
Merck
All Photos(1)

Key Documents

860688P

Avanti

14:0 SM (d18:1/14:0)

Avanti Research - A Croda Brand 860688P, powder

Synonym(s):

N-myristoyl-D-erythro-sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$875.00

$875.00


Available to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$875.00

About This Item

Empirical Formula (Hill Notation):
C37H75N2O6P
CAS Number:
Molecular Weight:
674.98
UNSPSC Code:
12352211
NACRES:
NA.25

$875.00


Available to ship on24 April 2025Details


Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860688P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860688P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C37H75N2O6P/c1-6-8-10-12-14-16-18-19-21-22-24-26-28-30-36(40)35(34-45-46(42,43)44-33-32-39(3,4)5)38-37(41)31-29-27-25-23-20-17-15-13-11-9-7-2/h28,30,35-36,40H,6-27,29,31-34H2,1-5H3,(H-,38,41,42,43)/b30-28+/t35-,36+/m0/s1

InChI key

KYICBZWZQPCUMO-PSALXKTOSA-N

General description

14:0 SM (d18:1/14:0), also known as N-myristoyl-D-erythro-sphingosylphosphorylcholine, is a sphingomyelin containing phosphocholine head group with ceramide (myristic acid (14:0) attached to sphingosine long chain base).[1] Sphingomyelins are mainly present in the outer leaflet of plasma ­membranes. They play a vital role in cell signaling.[2]

Application

N-myristoyl-D-erythro-sphingosylphosphorylcholine (14:0 SM (d18:1/14:0)) has been used as an analytical standard in ultra-high-performance liquid chromatography (UHPLC) coupled to high-resolution mass spectrometry (HRMS) for comprehensive polar lipidome characterization.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860688P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingomyelin and its role in cellular signaling
Chakraborty M and Jiang X
Lipidomics: Technologies and applications, 1-14 (2013)
Michela Antonelli et al.
Analytical and bioanalytical chemistry, 412(2), 413-423 (2019-11-25)
The chemical composition of Cannabis sativa L. has been extensively investigated for several years; nevertheless, a detailed lipidome characterization is completely lacking in the literature. To achieve this goal, an extraction and enrichment procedure was developed for the characterization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service