Skip to Content
Merck
All Photos(2)

Documents

131304P

Avanti

Glucosylceramide (Soy)

Avanti Research - A Croda Brand

Synonym(s):

Cerebroside (Soy); GlcCer(d18:2/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

Glucocerebrosides (Soy, 98%)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (131304P-10mg)
pkg of 1 × 25 mg (131304P-25mg)
pkg of 1 × 5 mg (131304P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

Glucosylceramide (GluCer) is a major sphingolipid of plant tissue and, thus, abundant in nature and in dietary food sources. This study characterized the molecular species of GluCer from soybean and wheat by low-resolution, high-resolution and tandem mass spectrometry. Soybean GluCer was comprised primarily (>98%) of ceramide with 4,8-sphingadiene (d18:2(delta4,delta8)) and alpha- hydroxypalmitic acid (h16:0); the remainder had the same backbone with h18:0, h20:0, h22:0 and h24:0 fatty acids. Wheat GluCer had three major ceramide, d18:2(delta4,delta8) with h16:0, d18:1(delta8) with h16:0 and d18: 2(delta4,delta8) with h20:0, and smaller amounts of other homologs. These backbones differ from those of mammalian sphingolipids, which often have a delta4-double bond (but rarely a delta8-double bond), and have alpha-hydroxy fatty acids in only some cases.

Application

Glucosylceramide (Soy) has been used for the quantitative analysis of plant sphingolipid long-chain bases by gas chromatography-mass spectrometry (GC-MS). It has also been used for enzyme linked immunosorbent assay (ELISA) assay.
Glucosylceramide (Soy) has been used:
  • in liposome preparation to test it effect on human α-synuclein aggregation
  • as a long chain base source to test alkaline hydrolysis
  • to test its immune modulatory effect on natural killer T lymphocytes and dendritic cells

Glucosylceramide (Soy) is suitable:
  • for intraperitoneal/oral administration, to treat mice in order to the study the period as an anti-inflammatory model to prevent fibrosis induction
  • for the preparation of glycolipids to study its effect on immune mediated colitis
  • to coat the wells of microtiter plate and perform enzyme linked immunosorbent assay (ELISA)

Biochem/physiol Actions

The lipid backbones of mammalian GluCer (sphingosine, d18:1(delta4), and ceramide) induce cell death (apoptosis) and inhibit colon carcinogenesis, it is critical to know the structures of GluCer present in plants as a first step toward understanding this potential link between diet and cancer. Previously unexplained fragmentations that were diagnostic for the type of sphingoid base backbone (i.e. by homolytic cleavage of the doubly allylic C-6-C-7 bond to yield a stable distonic allylic radical cation and an allylic radical neutral) were also identified. Hence this method should be useful in the identification of double bonds in sphingolipids, and structure-function relationships between sphingolipids and colon carcinogenesis.†

Packaging

5 mL Amber Glass Screw Cap Vial (131304P-10mg)
5 mL Amber Glass Screw Cap Vial (131304P-25mg)
5 mL Amber Glass Screw Cap Vial (131304P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amelioration of hepatic fibrosis via betaglucosylceramide-mediated immune modulation is associated with altered CD8 and NKT lymphocyte distribution
Safadi R, et al.
International Immunology, 19(8), 1021?1029-1021?1029 (2007)
Detection of antibody against fungal glucosylceramide in immunocompromised patients: a potential new diagnostic approach for cryptococcosis
Qureshi A, et al.
Mycopathologia, 173(5-6), 419-425 (2012)
Structure determination of soybean and wheat glucosylceramides by tandem mass spectrometry.
Sullards MC, et al.
Journal of Mass Spectrometry : Jms, 35, 347-353 (2000)
??Glucosylceramide: a novel method for enhancement of natural killer T lymphoycte plasticity in murine models of immune?mediated disorders
E Zigmond, et al.
Gut, 56(1), 82?89-82?89 (2007)
Detection of Antibody against Fungal Glucosylceramide in Immunocompromised Patients: A Potential New Diagnostic Approach for Cryptococcosis
Qureshi A, et al.
Mycopathologia, 173(5-6), 419?425-419?425 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service