Key Documents
Z690686
BactiZapper slide dryer attachment for 3 slides
aluminum holder (with a stainless steel clip)
Select a Size
1 EA
$122.00
$122.00
Check Cart for Availability
Select a Size
Change View
1 EA
$122.00
About This Item
UNSPSC Code:
41103200
NACRES:
NB.13
$122.00
Check Cart for Availability
material
aluminum holder (with a stainless steel clip)
manufacturer/tradename
BactiZapper
Benchmark Scientific B1000-03
General description
The BactiZapper slide dryer is an aluminum holder that attaches to the BactiZapper infrared sterilizer (Z690651, 230 V or Z690643, 115 V) to dry laboratory slides. The unit connects to the sterilizer with a stainless steel clip and uses heat radiating out of the device to dry laboratory slides.
The slide dryer holds three slides for drying multiple samples per use.
Legal Information
BactiZapper is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)