Z685445
Vacuubrand chemistry pumping units, manual control, Model PC 201 NT
AC/DC input 230 V AC, UK plug
Synonym(s):
Vacuubrand 20736403, diaphragm pump
Select a Size
$12,510.00
Estimated to ship on30 May 2025
Select a Size
About This Item
$12,510.00
Estimated to ship on30 May 2025
Recommended Products
manufacturer/tradename
Vacuubrand 737002
parameter
105 mbar vacuum
3.4-3.8 m3/h pumping speed
General description
The manual flow control valve regulates the effective pumping speed at the vacuum connection; the vacuum manometer offers an analog vacuum display. The separator at the inlet (AK), made of glass with a protective coating, retains particles and liquid droplets. The exhaust waste vapor condenser enables near-100-percent solvent recovery.
- Outstanding chemical resistance and superior vapor tolerance
- Exceptionally high performance even at low vacuum
- Excellent ultimate vacuum even with gas ballast
- Manual vacuum control, analog vacuum display
- Excellent environmental friendliness due to efficient solvent recovery
Supplied with MD 4C, manual flow control valve, analogue vacuum gauge, inlet catchpot and exhaust waste vapor condenser certification (NRTL).
Choose from one of the most recent versions:
Certificates of Analysis (COA)