Skip to Content
Merck
All Photos(3)

Key Documents

Z404586

IKA® ULTRA-TURRAX® disperser tools, stainless steel

dispersing element for T-25 digital disperser, 18 mm, 1/cs

Synonym(s):

disperser blades, tissue grinders, tissue homogenizers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$3,440.00

$3,440.00


Check Cart for Availability


Select a Size

Change View
1 EA
$3,440.00

About This Item

UNSPSC Code:
41103800
NACRES:
NB.22

$3,440.00


Check Cart for Availability

material

stainless steel

feature

stainless steel rotor/stator

packaging

pack of 1

manufacturer/tradename

IKA 0000593400

diam.

18 mm

Looking for similar products? Visit Product Comparison Guide

General description

The stainless steeldisperser tool IKA® ULTRA-TURRAX® has a shaft bearing of the dispersion toolsconsisting of a polytetrafluoroethylene (PTFE) slide bearing.

Application

Stainless steel disperser tool IKA® ULTRA-TURRAX® is suitable for emulsifying, dispersions, and wet crushing.

Legal Information

IKA is a registered trademark of IKA-Werke GmbH & Co. KG
ULTRA-TURRAX is a registered trademark of IKA-Werke GmbH & Co. KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service