Key Documents
Z309435
Quickfit® flat-bottom boiling flasks
capacity 500 mL, joint: ST/NS 24/29
Synonym(s):
FF500/3S
Select a Size
1 EA
$108.00
5 EA
$286.00
$108.00
Check Cart for Availability
Select a Size
Change View
1 EA
$108.00
5 EA
$286.00
About This Item
UNSPSC Code:
41121804
$108.00
Check Cart for Availability
material
borosilicate glass
flat bottom flask
short
single-neck
Joint
joint: ST/NS24/29
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 5
manufacturer/tradename
Quickfit FF 500/3S
availability
not available in USA and Canada
capacity
500 mL
General description
With short neck and ground-glass socket. Not suitable for vacuum use.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)