Key Documents
Z301329
Quickfit® plain bends
joint: ST/NS 24/29
Synonym(s):
RA9/33
Select a Size
1 EA
$111.00
$111.00
Check Cart for Availability
Select a Size
Change View
1 EA
$111.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$111.00
Check Cart for Availability
Joint
joint: ST/NS24/29
manufacturer/tradename
Quickfit RA 9/33
availability
not available in USA and Canada
not available in USA and Canada
General description
Socket to cone. Generally used as distilling adapters.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)