Key Documents
CLS70320250
Pyrex® VISTA round-bottom boiling flask
capacity 250 mL
Synonym(s):
boiling flask, bottom flask, round bottomed flask, Corning® 70320
Select a Size
2 EA
$98.30
4 EA
$105.00
$98.30
Available to ship on28 April 2025Details
Select a Size
Change View
2 EA
$98.30
4 EA
$105.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$98.30
Available to ship on28 April 2025Details
material
borosilicate glass
round bottom flask
short
Joint
joint ST/NS24/40
sterility
non-sterile
feature
closure type Standard taper joint
packaging
pack of 2
case of 4
manufacturer/tradename
Corning 70320-250
capacity
250 mL
neck size
(1)
General description
These round bottom boiling flasks have full length outer joints, but with shorter necks. Wall thicknesses are controlled to provide balance between thermal expansion and mechanical strength for greater shock resistance.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)