Key Documents
Z197130
Sigma-Aldrich
3M disposable respirator
organic vapor/acid gas, size L
Select a Size
1 EA
$51.50
$51.50
Check Cart for Availability
Select a Size
Change View
1 EA
$51.50
About This Item
UNSPSC Code:
46182002
NACRES:
NB.35
$51.50
Check Cart for Availability
size
L
packaging
pkg of 1 ea
manufacturer/tradename
3M 5000
General description
Pre-assembled disposable respirators. Low-profile design provides improved comfort and less vision interference. Approved for: Organic vapor/acid gas and dusts/mists having a TWA not less than 0.05 mg/m3 or 2 mppcf.
NIOSH approved.
Protection factor: Up to 10 × PEL.
Legal Information
3M is a trademark of 3M Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)