Skip to Content
Merck
All Photos(1)

Key Documents

22410-U

Sigma-Aldrich

Imp® tubing cutter

for use with 1/8 in. to 5/8 in. (4 to 15mm) O.D. tubing, pkg of 1 ea

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$88.30

$88.30


Available to ship on16 April 2025Details



Select a Size

Change View
1 EA
$88.30

About This Item

UNSPSC Code:
41122406
NACRES:
NB.35

$88.30


Available to ship on16 April 2025Details


packaging

pkg of 1 ea

manufacturer/tradename

Imp

tubing cutter L × W × D

1 11/16 in. × 1 5/16 in. × 7/8 in.

compatibility

for use with 1/8" to 5/8" (4 to 15mm O.D. tubing)

General description

Tool for cutting metal tubing.

Legal Information

Imp is a registered trademark of Stride Tool, Inc.

comparable product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service