Skip to Content
Merck
All Photos(1)

Key Documents

Z193275

Supelco

Sigma-Aldrich® TLC Plates

silica gel matrix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110504

material

aluminum support
silica gel matrix

feature

binder Polymeric
fluorescent indicator

packaging

pkg of 50 ea

manufacturer/tradename

Sigma-Aldrich

technique(s)

thin layer chromatography (TLC): suitable

L × W

5 cm × 10 cm

layer thickness

200 μm

particle size

25 μm

Looking for similar products? Visit Product Comparison Guide

General description

Thin-layer chromatography (TLC) plates for adsorption, partition, and ion-exchange techniques. Six media choices with glass, polyester, and aluminum backings. Silica-based adsorbent layers in Sigma-Aldrich TLC plates incorporate a polymeric binder; highly purified silica gel plates incorporate a gypsum/polymer binder.
  • Silica gel - for weakly to strongly polar separations, pore size 60 Å
  • High purity silica gel - acid washed for separation of aflatoxins
  • Cellulose - for partition chromatography, (SigmaCell Type 100 Cellulose)
  • Cellulose PEI - for separation of weak anions (amino acids, peptides) PEI plates may turn slightly yellow, affecting separation. Pre-develop plates in distilled water to minimize yellow color.
  • Chiral silica - for separation of optically active isomers by ligand exchange, pore size 60Å

Other Notes

this product will be discontinued, please find the alternative product here :
116834

Legal Information

SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service