Documents
Z192732
Horn tip for 500 and 750W ultrasonic processor
for horn 1/2 in.
About This Item
UNSPSC Code:
41101500
NACRES:
NB.77
manufacturer/tradename
Sonics & Materials 630-0406
Related Categories
General description
Tips screw into end of threaded horn. Replace when cavitation erosion becomes excessive.