Z183652
Poly-net® protective netting
blue, diam. 5/8-1 in., Close-mesh
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
Recommended Products
diam.
5/8-1 in.
thickness
0.07-1.2 mm , approximate
color
blue
Looking for similar products? Visit Product Comparison Guide
General description
PE net sleeve stretches around glassware to prevent scratching,chipping or breaking. Netting captures flying glass if container shatters. Easily cut to any length. Color-coded for size identification.
Application
Syringes and small bottles
Legal Information
Poly-net is a registered trademark of Norddeutsche Seekabelwerke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service