Skip to Content
Merck
All Photos(1)

Key Documents

CRMUMPLU

Lubricant; Multi-Parameter Certified Reference Material

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$959.00

$959.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
500 ML
$959.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$959.00


Available to ship on31 March 2025Details


Request a Bulk Order

grade

certified reference material

form

liquid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Asp. Tox. 1 - Carc. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service