Skip to Content
Merck
All Photos(1)

Key Documents

QBD10196

Sigma-Aldrich

Biotin-dPEG®11-NH2

Synonym(s):

Polyethylene glycol, alpha-(2-aminoethyl)-omega-[2-[[5-[(3aS,4S,6aR)-hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]ethoxy]-poly(oxy-1,2-ethanediyl)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$552.00
1000 MG
$2,010.00

$552.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$552.00
1000 MG
$2,010.00

About This Item

Empirical Formula (Hill Notation):
C34H66N4O13S
CAS Number:
Molecular Weight:
770.97
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

$552.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reaction type: Biotinylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(CCCCC1C2NC(NC2CS1)=O)=O

InChI key

SHPHAOSBFNWGAC-UHFFFAOYSA-N

Features and Benefits

Biotin-dPEG®11-amine is a single molecular weight dPEG® containing a terminal primary amine that permits biotin labeling with precise spacing to molecules containing a free carboxylic acid or aldehyde group. For small peptides and small molecules, the labeling can be site specific, while for larger molecules such as proteins, the labeling may be more random. The single molecular weight nature of the dPEG® spacer (44.1 Å) allows the user to precisely tailor the distance between the biotin and the target molecule in order to receive optimal binding between biotin and avidin.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service