Skip to Content
Merck
All Photos(1)

Key Documents

QBD11100

Sigma-Aldrich

Biotin-dPEG®3-oxyamine HCl

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$201.00

$201.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$201.00

About This Item

Empirical Formula (Hill Notation):
C18H35ClN4O6S
Molecular Weight:
471.01
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$201.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Biotinylations
reagent type: cross-linking reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(CCCCC1SCC(N2)C1NC2=O)NCCOCCOCCOCCO[NH3+].[Cl-]

InChI key

WWKHANWNJGBXKU-UHFFFAOYSA-N

Features and Benefits

Biotin-dPEG®3-oxyamine.HCl allows biotin labeling of molecules with free aldehydes or ketones. Biotin-dPEG®3-oxyamine-containing products forms oximes. These reagents are highly useful for labeling carbohydrates, glycoproteins, and other molecules with biotin following oxidation of the carbohydrate or glycan with sodium periodate or by chemical installation of an aldehyde or ketone on the target molecule. The dPEG® spacer is a single molecular weight PEG (16.6 Å) that provides excellent water solubility while extending the biotin moiety away from the surface of the labeled molecule for enhanced binding to avidin or streptavidin.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service