Skip to Content
Merck
All Photos(1)

Key Documents

QBD10193

Sigma-Aldrich

Biotin-dPEG®3-NH3+TFA-

>95% (HPLC)

Synonym(s):

Biotin-PEG-Amine, Biotin-PEG3-NH3-TFA-, Biotin-PEG3-amine TFA salt, Biotin-PEG3-ammonium trifluoroacetate

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$253.00

$253.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$253.00

About This Item

Empirical Formula (Hill Notation):
C22H39F3N4O7S
Molecular Weight:
560.63
UNSPSC Code:
12352200
NACRES:
NA.22

$253.00


Check Cart for Availability

Request a Bulk Order

Assay

>95% (HPLC)

form

solid or viscous liquid

reaction suitability

reaction type: Biotinylations
reaction type: Pegylations

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

Biotin-dPEG3-NH3+TFA- (Biotin-dPEG3-ammonium trifluoroacetate) reacts with activated esters (N-hydroxysuccinimide, 2,3,5,6-tetrafluorophenyl) of carboxylic acids under mildly basic conditions. It can be coupled directly to a carboxylic acid using 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide (EDC) chemistry. Also, the product can react with carbonyl groups (aldehydes or ketones) to form a Schiff base. The Schiff base can be reduced to a secondary amine using sodium cyanoborohydride if additional conjugate stability is necessary. Biotin-dPEG3-NH3+TFA- can be used to biotinylate antibodies useful in western blotting and immunohistochemical applications.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Legal Information

Products Protected under U.S. Patent #s 7,888,536 & 8,637,711 and European Patent #s 1,594,440 & 2,750,681
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service