Skip to Content
Merck
All Photos(1)

Documents

M62808

Sigma-Aldrich

4-Methyl-2-nitrophenol

99%

Synonym(s):

2-Nitro-p-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OH
CAS Number:
Molecular Weight:
153.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

bp

125 °C/22 mmHg (lit.)

mp

32-35 °C (lit.)

density

1.24 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(O)c(c1)[N+]([O-])=O

InChI

1S/C7H7NO3/c1-5-2-3-7(9)6(4-5)8(10)11/h2-4,9H,1H3

InChI key

SYDNSSSQVSOXTN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

226.4 °F - closed cup

Flash Point(C)

108 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hideo Kurebayashi et al.
Archives of toxicology, 76(12), 676-681 (2002-11-27)
Metabolism of 2-nitro- p-cresol (NPC), an important commercial chemical, was studied in female Sprague-Dawley rats, using (14)C-NPC. It was found that NPC was rapidly absorbed and excreted after an oral dose of 250 mg/kg. Approximately 90% of the administered dose
[Hepatic ornithine decarboxylase activity of rats induced by administration of 2-nitro-p-cresol and sodium deoxycholate].
S Nambaru et al.
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences, (106)(106), 95-97 (1988-01-01)
Jamshid Behin et al.
Water research, 121, 120-128 (2017-05-20)
In present work, the treatment of aromatic compounds of simulated wastewater was performed by Fenton and NaOCl/Fe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service