Skip to Content
Merck
All Photos(1)

Key Documents

T28517

Sigma-Aldrich

3-Methylbenzenethiol

95%

Synonym(s):

m-Thiocresol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$109.00

$109.00


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
5 G
$109.00

About This Item

Linear Formula:
CH3C6H4SH
CAS Number:
Molecular Weight:
124.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
95%

$109.00


Estimated to ship on28 March 2025


Request a Bulk Order

Quality Level

Assay

95%

refractive index

n20/D 1.5762 (lit.)

bp

196 °C (lit.)

density

1.044 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(S)c1

InChI

1S/C7H8S/c1-6-3-2-4-7(8)5-6/h2-5,8H,1H3

InChI key

WRXOZRLZDJAYDR-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Javier Campanini-Salinas et al.
Molecules (Basel, Switzerland), 23(7) (2018-07-22)
A rapid emergence of resistant bacteria is occurring worldwide, endangering the efficacy of antibiotics and reducing the therapeutic arsenal available for treatment of infectious diseases. In the present study, we developed a new class of compounds with antibacterial activity obtained

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service