Skip to Content
Merck
All Photos(1)

Key Documents

F15315

Sigma-Aldrich

4-Fluorothiophenol

98%

Synonym(s):

4-Fluorobenzenethiol, 4-Mercaptofluorobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$116.00
25 G
$120.00

$116.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$116.00
25 G
$120.00

About This Item

Linear Formula:
FC6H4SH
CAS Number:
Molecular Weight:
128.17
Beilstein:
1446350
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$116.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.550 (lit.)

density

1.203 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(S)cc1

InChI

1S/C6H5FS/c7-5-1-3-6(8)4-2-5/h1-4,8H

InChI key

OKIHXNKYYGUVTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Krishnadas et al.
ACS nano, 11(6), 6015-6023 (2017-05-18)
Monolayer protected clusters exhibit rich diversity in geometric and electronic structures. However, structure-reactivity relationships in these clusters are rarely explored. In this context, [Ag
Homan Kang et al.
International journal of molecular sciences, 22(4) (2021-02-14)
We present a template-assisted method for synthesizing nanogap shell structures for biomolecular detections based on surface-enhanced Raman scattering. The interior nanogap-containing a silver shell structure, referred to as a silver nanogap shell (Ag NGS), was fabricated on silver nanoparticles (Ag
Hyung-Mo Kim et al.
Scientific reports, 8(1), 13938-13938 (2018-09-19)
In this study, we report on the fabrication of multilayered tri-functional magnetic-SERS-fluorescence nanoprobes (MF-SERS particles) containing clustered superparamagnetic Fe3O4 nanoparticles (NPs), silver NPs, and a fluorescent silica layer. The MF-SERS particles exhibited strong SERS signals from the silver NPs as
Myeong Geun Cha et al.
Nanoscale, 9(34), 12556-12564 (2017-08-19)
Multimodal imaging can provide complementary biomedical information which has huge potential in pre-clinical and clinical imaging and sensing. In this study, we introduce dual modal NIR silver bumpy nanoprobes for in vivo imaging and multiplexed detection of biomolecules by both
Esma Khatun et al.
Nanoscale, 9(24), 8240-8248 (2017-06-06)
We report the synthesis of a new silver cluster, [Ag

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service