Skip to Content
Merck
All Photos(4)

Key Documents

M1875

Sigma-Aldrich

Sodium malonate dibasic monohydrate

≥98% (non-aqueous titration)

Synonym(s):

Malonic acid disodium salt monohydrate, Propanedioic acid disodium salt monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$100.00
100 G
$189.00
500 G
$751.00

$100.00


Available to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$100.00
100 G
$189.00
500 G
$751.00

About This Item

Linear Formula:
CH2(CO2Na)2 · H2O
CAS Number:
Molecular Weight:
166.04
Beilstein:
3772420
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$100.00


Available to ship on17 April 2025Details


Request a Bulk Order

Assay

≥98% (non-aqueous titration)

SMILES string

O.[Na].OC(=O)CC(O)=O

InChI

1S/C3H4O4.2Na/c4-2(5)1-3(6)7;;/h1H2,(H,4,5)(H,6,7);;/q;2*+1/p-2

InChI key

PRWXGRGLHYDWPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium malonate dibasic monohydrate, also known as Malonic acid disodium salt monohydrate, is commonly used as a building block in organic reaction for synthesizing complex compounds.

Application

Sodium malonate dibasic monohydrate can be used as a ligand to synthesize:
  • Bis(cyclopentadienyl)molybdenum malonate using molybdenocene dichloride in the presence of water[1].
  • Malonato complexes of manganese using manganese dichloride and 1,10-phenanthroline in the presence of methanol[2].
  • Metal-organic based polymeric complex using transition metal compound and urotropine[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjit Konar et al.
Inorganic chemistry, 42(8), 2545-2552 (2003-04-15)
Two new metal-organic based polymeric complexes, [Cu(4)(O(2)CCH(2)CO(2))(4)(L)].7H(2)O (1) and [Co(2)(O(2)CCH(2)CO(2))(2)(L)].2H(2)O (2) [L = hexamethylenetetramine (urotropine)], have been synthesized and characterized by X-ray crystal structure determination and magnetic studies. Complex 1 is a 1D coordination polymer comprising a carboxylato bridged Cu(4)
Idainés Feliciano et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 14(7), 1109-1117 (2009-06-19)
Two water-soluble molybdenocene complexes containing oxygen chelating ligands, maltolato and malonate, have been synthesized to elucidate the role of the ancillary ligands in the molybdenocene cytotoxic activity. The structural characterizations of these species by 1H NMR and IR spectroscopies suggest
Two new supramolecular malonato complexes of manganese (II): synthesis, crystal structure and magnetic property
Sain S, et al.
Inorganica chimica acta, 351, 12-20 (2003)
Milya Davlieva et al.
Nucleic acids research, 43(9), 4758-4773 (2015-04-22)
LiaR is a 'master regulator' of the cell envelope stress response in enterococci and many other Gram-positive organisms. Mutations to liaR can lead to antibiotic resistance to a variety of antibiotics including the cyclic lipopeptide daptomycin. LiaR is phosphorylated in
Alessio Innocenti et al.
Bioorganic & medicinal chemistry letters, 15(3), 573-578 (2005-01-25)
A detailed inhibition study of five carbonic anhydrase (CA, EC 4.2.1.1) isozymes with carboxylates including aliphatic (formate, acetate), dicarboxylic (oxalate, malonate), hydroxy/keto acids (l-lactate, l-malate, pyruvate), tricarboxylic (citrate), or aromatic (benzoate, tetrafluorobenzoate) representatives, some of which are important intermediates in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service