Skip to Content
Merck
Sodium malonate dibasic monohydrate BioXtra

Sodium malonate dibasic monohydrate

Sodium malonate dibasic monohydrate BioXtra
Synonym(s):
Malonic acid disodium salt monohydrate, Propanedioic acid disodium salt monohydrate
Linear Formula:
CH2(CO2Na)2 · H2O
CAS Number:
Molecular Weight:
166.04
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing