Key Documents
HAM240134
Dispense tubing for Hamilton® Microlab® 600 pipettor
FEP, gauge × L 18 × 1372 mm
Select a Size
1 EA
$107.00
$107.00
Check Cart for Availability
Select a Size
Change View
1 EA
$107.00
About This Item
UNSPSC Code:
41105108
NACRES:
NB.01
$107.00
Check Cart for Availability
material
FEP
gauge × L
18 × 1372 mm
fitting
1/4-28 tubing tapered
suitability
suitable for
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)