Skip to Content
Merck
All Photos(1)

Key Documents

DWKWGW447P

Azlon Square Shoulder Unvented Wash Bottles

wide-neck, low-density polyethylene bottle, green polypropylene closure, capacity 500 mL

Synonym(s):

WGW447P

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$88.20

$88.20


Check Cart for Availability


Select a Size

Change View
5 EA
$88.20

About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

$88.20


Check Cart for Availability

material

green polypropylene closure
low-density polyethylene bottle
wide-neck

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGW447P

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for solvent and water dispensing within the laboratory

Features and Benefits

Patented Driplok vapour venting valve to prevent solvent drips

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service