CLS694217L
Pyrex® cylindrical jar, plain
O.D. × H 305 mm × 305 mm
Synonym(s):
lab jar, lab jars, laboratory jars, Corning® 6942
About This Item
Recommended Products
material
borosilicate glass bottle
clear bottle
cylindrical bottle
feature
cap: no
packaging
case of 1 ea
pack of 1 ea
manufacturer/tradename
Corning 6942-17L
availability
not available in EU (Not available in EU countries)
O.D. × H
305 mm × 305 mm
bottle neck diam.
305 mm
capacity
17.0 L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service