Skip to Content
Merck
Pyrex® cylindrical jar, plain O.D. × H 406 mm × 305 mm

Pyrex® cylindrical jar, plain

Pyrex® cylindrical jar, plain O.D. × H 406 mm × 305 mm
Synonym(s):
Corning® 6942

Products

Manufacturer
Capacity (L)
Diameter (mm)
OD (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size