CLS66402125
PYREXPLUS® Squibb separatory funnel, Pyrex® tapered stopper, PTFE stopcock
funnel capacity 125 mL
Synonym(s):
Corning® 66402
Select a Size
$503.00
Select a Size
About This Item
$503.00
Recommended Products
stopcock size
2
stopper size
22
manufacturer/tradename
Corning 66402-125
funnel capacity
125 mL
stem O.D. × L
10 mm × 65 mm , below stopcock
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service