Skip to Content
Merck
All Photos(1)

Key Documents

64805-U

Supelco

Separatory Funnel

PTFE stopcock, size 250 mL

Synonym(s):

SEPARATORY FUNNEL 250ML W/PTFE STOPCOCK, EA

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$210.00

$210.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$210.00

About This Item

UNSPSC Code:
41121809
NACRES:
NB.21

$210.00


Check Cart for Availability

Request a Bulk Order

material

PTFE stopcock

size

22 (stopper)
4 (stopcock)
250 mL

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

General description

Squibb-type, with glass or PTFE stopcock plug.

Application

  • DAPNe with micro-capillary separatory chemistry-coupled to MALDI-MS for the analysis of polar and non-polar lipid metabolism in one cell.: This study introduces a novel approach combining micro-capillary separatory chemistry with MALDI-MS to analyze lipid metabolism within a single cell. The technique allows for the separation and analysis of both polar and non-polar lipids, providing comprehensive insights into cellular lipidomics (Hamilton et al., 2017).
  • Reverse phase liquid chromatographic determination of sulfathiazole residues in honey.: This research demonstrates the use of reverse phase liquid chromatography in conjunction with a separatory funnel to detect and quantify sulfathiazole residues in honey. The method provides a reliable and efficient approach to ensuring food safety and regulatory compliance (Barry and MacEachern, 1983).
  • Economic method for analysis of fluid milk for organochlorine residues at the 10 ppb level.: The study outlines an economical method for analyzing organochlorine pesticide residues in milk using a separatory funnel. This technique achieves detection limits at the 10 ppb level, making it a valuable tool for food quality control (Veierov and Aharonson, 1980).

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service