CLS4985500
Pyrex® narrow-mouth Erlenmeyer flask with screw-cap
capacity 500 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 4985
Select a Size
$925.00
Select a Size
About This Item
$925.00
Recommended Products
material
borosilicate glass
narrow-mouth
phenolic cap
screw top cap
neck thread size
38-430
sterility
non-sterile
feature
cap
closure type Screw cap
subdivision 50 mL
packaging
case of 24
pack of 6
manufacturer/tradename
Corning 4985-500
volume measuring range
100-500 mL
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service