CLS36060350F
Pyrex® Buchner funnel with fritted disc
capacity 350 mL, porosity 4-5.5 μm
Synonym(s):
Corning® 36060
Select a Size
$507.00
Available to ship on23 April 2025Details
Select a Size
About This Item
$507.00
Available to ship on23 April 2025Details
Recommended Products
manufacturer/tradename
Corning 36060-350F
capacity
350 mL
disc diam.
80 mm
pore size
4-5.5 μm porosity
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)