Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
Buchner funnel with fritted disc
Synonym(s):
Corning
®
36060
Products
Product Category
funnels (24)
Manufacturer
Capacity (mL)
Diameter (mm)
Pore size (µm)
Filter Products
Showing 1-20 of 24
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CLS360602LM
Pyrex
®
Buchner funnel with fritted disc,
capacity 2,000 mL, porosity 10-15 μm
Expand
View Pricing
CLS360603LM
Pyrex
®
Buchner funnel with fritted disc,
capacity 3,000 mL, porosity 10-15 μm
Expand
View Pricing
CLS36060600M
Pyrex
®
Buchner funnel with fritted disc,
capacity 600 mL, porosity 10-15 μm
Expand
View Pricing
CLS3606060C
Pyrex
®
Buchner funnel with fritted disc,
capacity 60 mL, porosity 40-60 μm
Expand
View Pricing
CLS3606060F
Pyrex
®
Buchner funnel with fritted disc,
capacity 60 mL, porosity 4-5.5 μm
Expand
View Pricing
CLS360603LC
Pyrex
®
Buchner funnel with fritted disc,
capacity 3,000 mL, porosity 40-60 μm
Expand
View Pricing
CLS3606030M
Pyrex
®
Buchner funnel with fritted disc,
capacity 30 mL, porosity 10-15 μm
Expand
View Pricing
CLS36060150F
Pyrex
®
Buchner funnel with fritted disc,
capacity 150 mL, porosity 4-5.5 μm
Expand
View Pricing
CLS3606015F
Pyrex
®
Buchner funnel with fritted disc,
capacity 15 mL, porosity 4-5.5 μm
Expand
View Pricing
CLS36060600F
Pyrex
®
Buchner funnel with fritted disc,
capacity 600 mL, porosity 4-5.5 μm
Expand
View Pricing
CLS36060150M
Pyrex
®
Buchner funnel with fritted disc,
capacity 150 mL, porosity 10-15 μm
Expand
View Pricing
CLS360602F
Pyrex
®
Buchner funnel with fritted disc,
capacity 2 mL
Expand
View Pricing
CLS36060350F
Pyrex
®
Buchner funnel with fritted disc,
capacity 350 mL, porosity 4-5.5 μm
Expand
View Pricing
CLS3606015C
Pyrex
®
Buchner funnel with fritted disc,
capacity 15 mL, porosity 40-60 μm
Expand
View Pricing
CLS3606030C
Pyrex
®
Buchner funnel with fritted disc,
capacity 30 mL, porosity 40-60 μm
Expand
View Pricing
CLS3606030F
Pyrex
®
Buchner funnel with fritted disc,
capacity 30 mL, porosity 4-5.5 μm
Expand
View Pricing
CLS36060350C
Pyrex
®
Buchner funnel with fritted disc,
capacity 350 mL, porosity 40-60 μm
Expand
View Pricing
CLS3606060M
Pyrex
®
Buchner funnel with fritted disc,
capacity 60 mL, porosity 10-15 μm
Expand
View Pricing
CLS36060350M
Pyrex
®
Buchner funnel with fritted disc,
capacity 350 mL, porosity 10-15 μm
Expand
View Pricing
CLS36060600C
Pyrex
®
Buchner funnel with fritted disc,
capacity 600 mL, porosity 40-60 μm
Expand
View Pricing
Page 1 of 2
Page 1 of 2