CLS360602F
Pyrex® Buchner funnel with fritted disc
capacity 2 mL
Synonym(s):
Corning® 36060
Select a Size
$81.30
Select a Size
About This Item
$81.30
Recommended Products
packaging
case of 1
manufacturer/tradename
Corning 3606-02F
H
70 mm
capacity
2 mL
disc diam.
10 mm
pore size
4-5.5 μm pore size
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service