BR88635
BRAND® stopcock one-way with nozzles
for tube I.D. 8 mm
Synonym(s):
PTFE, Stopcocks
All Photos(1)
About This Item
Recommended Products
material
PTFE
packaging
pack of 1 ea
manufacturer/tradename
BRAND 88635
I.D.
4 mm
Related Categories
General description
The BRAND® one-way stopcock with nozzles is constructed from high-quality PTFE, offering outstanding chemical resistance for various laboratory applications. This stopcock features a valve lever made of polypropylene (PP) for easy operation and control. It is equipped with integrated hose nozzles, facilitating efficient fluid transfer and connection to hoses. The robust design ensures reliable performance, making it suitable for handling aggressive chemicals and solvents.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG