Documents
BAF132050002
SP Bel-Art® Benchtop Biohazard Incinerator Cartons
width × H × L 196 mm (8 in.) × 10 in. (254 mm) × 8 in. (196 mm)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
manufacturer/tradename
Bel-Art F13205-0002
width × H × L
196 mm (8 in.) × 10 in. (254 mm) × 8 in. (196 mm)
General description
This corrugated cardboard receptacle is perfect for containing biohazardous waste slated for incineration. When the box is full, pull the flap to a closed position and the entire unit can be placed in the incinerator.
Features and Benefits
  • Tabs along the top edge keep a clear 0.038mm (1.5 mil) thick polyethylene biohazard bag from slipping out of place
  • Snug-fitting lid has a flap for easily inserting waste material
  • Printed with large, bright, biohazard graphics and instructions in four languages (English, Spanish, French, German)
  • Packaged flat to save space and shipping costs; carton and inner bag can be assembled in seconds
  • Available in sizes to suit benchtop or floor use
  • Optional, reusable, sanitary cover (F13204-0000) for floor model sold separately
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP