Skip to Content
Merck
All Photos(2)

Key Documents

BAF131970000

SP Bel-Art® Biohazard Disposal Can

polypropylene

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$114.00

$114.00


Check Cart for Availability


Select a Size

Change View
1 EACH
$114.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.43

$114.00


Check Cart for Availability

material

polypropylene

manufacturer/tradename

Bel-Art F13197-0000

width × H × L

305 mm (12 in.) × 11.5 in. (290 mm) × 9 in. (225 mm)

General description

Bright red safety disposal container is suitable for biological/biohazard materials, broken glass, recyclables or other waste and sized for benchtop or floor placement.

Features and Benefits

Bright red safety disposal container is suitable for biological/biohazard materials, broken glass, recyclables or other waste and sized for benchtop or floor placement.
  • One piece hinged lid protects users from contact with contents; helps retain odors and contain airborne contaminants
  • Molded of polypropylene for long service life
  • Puncture-resistant; autoclavable at 121°C (250°F)
  • 19 x 23" autoclave bags (F13164-1923 or F13165-1923) fit perfectly and are secured with an internal stainless steel wire bar

Legal Information

Bel-Art is a registered trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service