Select a Size
$152.00
Select a Size
About This Item
$152.00
Recommended Products
material
red polypropylene
feature
Holds sharps
packaging
pack of 18 ea
manufacturer/tradename
Heathrow Scientific HS120177
size
115x115x192 mm
capacity
1 lb
Related Categories
General description
- Stackable to reduce storage space before use
- 1.0 L capacity (Quart) with a clear lid to view fill level
- Slide access lid for temporary and final closures for safety and security
- Built-in needle removal port, preventing direct contact with sharps
- Compatible with the Droplet Blood Collection Tray
- Measures 4.5 x 4.5 x 7.5 in (11.5 x 11.5 x 19.2 cm)
- Pack of 18
accessory
Choose from one of the most recent versions:
Certificates of Analysis (COA)