Skip to Content
Merck
All Photos(1)

Key Documents

B67007

Sigma-Aldrich

1-Bromo-3-fluorobenzene

≥99%

Synonym(s):

1-Fluoro-3-bromobenzene, 3-Bromo-1-fluorobenzene, m-Fluorophenyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$38.50
25 G
$50.90
100 G
$105.00

$38.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$38.50
25 G
$50.90
100 G
$105.00

About This Item

Linear Formula:
BrC6H4F
CAS Number:
Molecular Weight:
175.00
Beilstein:
1853969
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$38.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.526 (lit.)

bp

149-151 °C (lit.)

density

1.567 g/mL at 25 °C (lit.)

SMILES string

Fc1cccc(Br)c1

InChI

1S/C6H4BrF/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

QDFKKJYEIFBEFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

100.4 °F

Flash Point(C)

38 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hannah Watson et al.
The Journal of experimental biology, 221(Pt 13) (2018-05-12)
In oviparous animals, maternally transferred antioxidants protect the embryo from oxidative damage from high rates of reactive oxygen species (ROS) production incurred by rapid growth. Elevated ROS exposure, beyond that incurred by normal growth, can occur as a result of
Cas Eikenaar et al.
Physiological and biochemical zoology : PBZ, 90(2), 223-229 (2017-03-10)
Most avian migrants alternate flight bouts, characterized by high metabolic rates, with stopovers, periods of fuel replenishment through hyperphagia. High-energy metabolism and excessive calorie intake shift the balance between damaging prooxidants and antioxidants toward the former. Hence, migration likely affects
Shigeo Hayashi et al.
Journal of enzyme inhibition and medicinal chemistry, 29(6), 846-867 (2014-02-13)
Because of the pivotal role of cyclooxygenase (COX) in the inflammatory processes, non-steroidal anti-inflammatory drugs (NSAIDs) that suppress COX activities have been used clinically for the treatment of inflammatory diseases/syndromes; however, traditional NSAIDs exhibit serious side-effects such as gastrointestinal damage

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service