Skip to Content
Merck
All Photos(1)

Key Documents

B66809

Sigma-Aldrich

1-Bromo-2-fluorobenzene

99%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$92.30
100 G
$210.00

$92.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$92.30
100 G
$210.00

About This Item

Linear Formula:
BrC6H4F
CAS Number:
Molecular Weight:
175.00
Beilstein:
1071819
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$92.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.534 (lit.)

density

1.601 g/mL at 25 °C (lit.)

SMILES string

Fc1ccccc1Br

InChI

1S/C6H4BrF/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

IPWBFGUBXWMIPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen J Fowler et al.
Thorax, 70(4), 320-325 (2015-02-11)
Healthcare associated infections, including ventilator associated pneumonia, are difficult to diagnose and treat, and are associated with significant morbidity, mortality and cost. We aimed to demonstrate proof of concept that breath volatile profiles were associated with the presence of clinically

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service