Skip to Content
Merck
All Photos(2)

Key Documents

550671

Sigma-Aldrich

3-Fluorophenylmagnesium bromide solution

1.0 M in THF

Synonym(s):

m-Fluorophenylmagnesium bromide, Bromo(3-fluorophenyl)magnesium

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$249.00
4 X 25 ML
$260.00

$249.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
$249.00
4 X 25 ML
$260.00

About This Item

Linear Formula:
FC6H4MgBr
CAS Number:
Molecular Weight:
199.30
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$249.00


Check Cart for Availability

Request a Bulk Order

reaction suitability

reaction type: Grignard Reaction

Quality Level

concentration

1.0 M in THF

bp

65 °C

density

1.024 g/mL at 25 °C

functional group

fluoro

SMILES string

Fc1cccc([Mg]Br)c1

InChI

1S/C6H4F.BrH.Mg/c7-6-4-2-1-3-5-6;;/h1-2,4-5H;1H;/q;;+1/p-1

InChI key

VFPDAAQAKDGSHQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.0 °F - closed cup

Flash Point(C)

-21.11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service