Skip to Content
Merck
All Photos(2)

Key Documents

B59209

Sigma-Aldrich

4-Bromobenzoyl chloride

98%

Synonym(s):

4-Bromo-1-benzenecarbonyl chloride, p-Bromobenzoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$62.90
25 G
$129.00

$62.90


Estimated to ship on29 March 2025


Request a Bulk Order

Select a Size

Change View
10 G
$62.90
25 G
$129.00

About This Item

Linear Formula:
BrC6H4COCl
CAS Number:
Molecular Weight:
219.46
Beilstein:
636641
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
crystals
Assay:
98%

$62.90


Estimated to ship on29 March 2025


Request a Bulk Order

Quality Level

Assay

98%

form

crystals

bp

174 °C/102 mmHg (lit.)

mp

36-39 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(=O)c1ccc(Br)cc1

InChI

1S/C7H4BrClO/c8-6-3-1-5(2-4-6)7(9)10/h1-4H

InChI key

DENKGPBHLYFNGK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Miyagaki et al.
Langmuir : the ACS journal of surfaces and colloids, 35(30), 9761-9768 (2019-06-15)
Poly(ether ether ketone) (PEEK) possesses attractive mechanical and thermal properties but demonstrates poor adhesion. To overcome this disadvantage, in this study, the surface modification of PEEK or PEEK-based carbon-fiber-reinforced thermoplastics (CFRTP) was performed through the Friedel-Crafts reaction and successive epoxidation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service