Skip to Content
Merck
All Photos(1)

Key Documents

C26801

Sigma-Aldrich

3-Chlorobenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$71.40
100 G
$135.00

$71.40


Estimated to ship on28 March 2025


Request a Bulk Order

Select a Size

Change View
25 G
$71.40
100 G
$135.00

About This Item

Linear Formula:
ClC6H4COCl
CAS Number:
Molecular Weight:
175.01
Beilstein:
386256
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

$71.40


Estimated to ship on28 March 2025


Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.569 (lit.)

bp

225 °C (lit.)

density

1.367 g/mL at 20 °C (lit.)

SMILES string

ClC(=O)c1cccc(Cl)c1

InChI

1S/C7H4Cl2O/c8-6-3-1-2-5(4-6)7(9)10/h1-4H

InChI key

WHIHIKVIWVIIER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hina Siddiqui et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 13(5), 465-476 (2017-01-27)
Multidrug resistance (MDR) is a major challenge in the treatment of infectious diseases. The MDR in urinary tract infection causing bacteria, such as Escherichia coli, has made treatment of UTI very difficult. The aims of the current study were to
Dongsheng Xu et al.
Journal of chromatography. A, 1593, 63-72 (2019-02-05)
An O-[2-(methacryloyloxy)-ethylcarbamoyl]-10,11-dihydroquinidine (MQD)-silica hybrid monolithic column was prepared by a facile "one-step" strategy within a 100 μm I.D. capillary. The influence of the methanol, ethylene glycol and water volume ratio, reaction temperature and time, cetyltrimethylammonium bromide and MQD monomers content
Ataf Ali Altaf et al.
Dalton transactions (Cambridge, England : 2003), 47(34), 11769-11781 (2018-08-18)
Twenty-three compounds in two series of ferrocene-based anilides, with the general formula C5H5-Fe-C5H4-C6H4-NH-CO-C6H4-R (where R = H, F, Cl, CH3 and OCH3), have been successfully synthesized. The compounds were characterized by elemental analysis and FTIR, 1H NMR and 13C NMR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service