Skip to Content
Merck
All Photos(2)

Key Documents

230243

Sigma-Aldrich

3-Methoxybenzoyl chloride

99%

Synonym(s):

m-Anisoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$63.20
25 G
$219.00

$63.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$63.20
25 G
$219.00

About This Item

Linear Formula:
CH3OC6H4COCl
CAS Number:
Molecular Weight:
170.59
Beilstein:
386659
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$63.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.558 (lit.)

bp

123-125 °C/15 mmHg (lit.)

density

1.214 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

COc1cccc(c1)C(Cl)=O

InChI

1S/C8H7ClO2/c1-11-7-4-2-3-6(5-7)8(9)10/h2-5H,1H3

InChI key

RUQIUASLAXJZIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ataf Ali Altaf et al.
Dalton transactions (Cambridge, England : 2003), 47(34), 11769-11781 (2018-08-18)
Twenty-three compounds in two series of ferrocene-based anilides, with the general formula C5H5-Fe-C5H4-C6H4-NH-CO-C6H4-R (where R = H, F, Cl, CH3 and OCH3), have been successfully synthesized. The compounds were characterized by elemental analysis and FTIR, 1H NMR and 13C NMR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service