Skip to Content
Merck
All Photos(3)

Key Documents

ALD00574

Sigma-Aldrich

(1R,4R)-1-((4-(Trifluoromethyl)phenyl)sulfonyl)bicyclo[2.1.0]pentane, (-)-R-enantiomer

95%

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$285.00

$285.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
$285.00

About This Item

Empirical Formula (Hill Notation):
C12H11F3O2S
CAS Number:
Molecular Weight:
276.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$285.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

powder or chunks (crystals)

reaction suitability

reaction type: C-C Bond Formation

SMILES string

FC(F)(F)c1ccc(cc1)[S](=O)(=O)C32C(C3)CC2

InChI key

KAKTWVYNDZQZCE-UHFFFAOYSA-N

Application

The following "spring-loaded" electrophile is a convenient reagent for the installation of cyclopentane to primary and secondary amines. This product is one of several synthetically useful reagents reported by Baran and coworkers for the “any-stage” installation of small, strained ring systems.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service