Skip to Content
Merck
All Photos(1)

Key Documents

ALD00468

Sigma-Aldrich

sodium (4-chlorophenyl)methanesulfinate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$415.00

$415.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$415.00

About This Item

Empirical Formula (Hill Notation):
C7H6ClNaO2S
Molecular Weight:
212.63
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.06

$415.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

reaction suitability

reaction type: C-C Bond Formation

SMILES string

ClC1=CC=C(CS(O[Na])=O)C=C1

InChI

1S/C7H7ClO2S.Na/c8-7-3-1-6(2-4-7)5-11(9)10;/h1-4H,5H2,(H,9,10);/q;+1/p-1

InChI key

UXCLCCYBFMAOBU-UHFFFAOYSA-M

General description

Sodium (4-chlorophenyl)methanesulfinate is an alkyl Baran Diversinate. These reagents facilitate the functionalization of heteroaromatic C-H bonds under simple reaction conditions.

Application

The following Baran Sulfinate is a part of a toolbox of diversification reagents, which are ideal for the late-stage functionalization of nitrogen-containing heterocycles. The regioselectivity can be effectively tuned by modification of the pH and solvent selection.

Legal Information

Diversinates is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service