Skip to Content
Merck
All Photos(1)

Key Documents

900258

Sigma-Aldrich

Isopropoxy(phenyl)silane

Synonym(s):

RubenSilane

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$230.00
5 G
$776.00

$230.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$230.00
5 G
$776.00

About This Item

Empirical Formula (Hill Notation):
C9H14OSi
CAS Number:
Molecular Weight:
166.29
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

$230.00


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

reaction suitability

reagent type: reductant
reaction type: Reductions

refractive index

n/D 1.4799

density

0.926 g/mL

storage temp.

2-8°C

SMILES string

[SiH2](OC(C)C)c1ccccc1

InChI

1S/C9H14OSi/c1-8(2)10-11-9-6-4-3-5-7-9/h3-8H,11H2,1-2H3

InChI key

XYMAHMLQCLMTDN-UHFFFAOYSA-N

General description

Isopropoxy(phenyl)silane is used as a stoichiometric reductant that allows for a significant decrease in catalyst loading, lower reaction temperatures, a wide range of functional group tolerance, and more diverse solvents in metal-catalyzed Mukaiyama hydrofunctionalizations.

Application

Isopropoxy(phenyl)silane can be used as a hydride source for alkene hydrofunctionalization reactions in the presence of metal catalysts, particularly under aprotic, nonalcoholic conditions. This reagent is considered as one of the most efficient stoichiometric reductants than phenylsilane because of its ability to exclude alcohol solvent from a series of catalytic reactions. When utilized, it allows the researcher to significantly decrease catalyst loadings, lower reaction temperatures, and employ more diverse solvents in iron- and manganese-catalyzed Mukaiyama hydrofunctionalizations.[1][2]
It can be used as a reagent in:
  • The hydrogenation of olefins
  • Branch-selective olefin cross-coupling reactions
  • Markovnikov alkene hydration/amination reactions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isopropoxy (phenyl) silane
Demoret RM and Shenvi RA
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Carla Obradors et al.
Journal of the American Chemical Society, 138(14), 4962-4971 (2016-03-18)
We report the discovery of an outstanding reductant for metal-catalyzed radical hydrofunctionalization reactions. Observations of unexpected silane solvolysis distributions in the HAT-initiated hydrogenation of alkenes reveal that phenylsilane is not the kinetically preferred reductant in many of these transformations. Instead

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service