Skip to Content
Merck
All Photos(3)

Key Documents

A30702

Sigma-Aldrich

Allyl chloride

reagent grade, 98%

Synonym(s):

3-Chloro-1-propene, Chlorallylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2Cl
CAS Number:
Molecular Weight:
76.52
Beilstein:
635704
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

grade

reagent grade

Quality Level

vapor density

2.6 (vs air)

vapor pressure

20.58 psi ( 55 °C)
5.71 psi ( 20 °C)

Assay

98%

form

liquid

expl. lim.

11.2 %

refractive index

n20/D 1.414 (lit.)

bp

44-46 °C (lit.)

mp

−130 °C (lit.)

density

0.939 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

ClCC=C

InChI

1S/C3H5Cl/c1-2-3-4/h2H,1,3H2

InChI key

OSDWBNJEKMUWAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Allyl chloride can be used as a reactant to prepare:
  • 1-Allyl-2-alkynylbenzene derivatives through allylalkynylation of in situ generated benzynes.
  • α,ω-Difunctional intermediates by reacting with fatty acid methyl esters using ruthenium catalysts.
  • Homoallylic alcohols through Sn/I2 catalyzed Barbier-type allylation reactions of carbonyl compounds.

Other Notes

The article number A30702-4X1L will be discontinued. Please order the single bottle A30702-1L which is physically identical with the same exact specifications.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Nervous system,Liver,Kidney, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-25.6 °F

Flash Point(C)

-32 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Airong Xu et al.
Polymers, 11(5) (2019-05-15)
Cellulose is the most abundant natural biopolymer, with unique properties such as biodegradability, biocompability, nontoxicity, and so on. However, its extensive application has actually been hindered, because of its insolubility in water and most solvents. Herein, highly efficient cellulose solvents
Sn/I2 Mediated allylation of carbonyl compounds with allyl (crotyl) halide in water
Zhou Y, et al.
ARKIVOC (Gainesville, FL, United States), 11(1), 142-153 (2008)
Palladium-Catalyzed Allylalkynylation of Benzynes: A Highly Efficient Route to Substituted 1-Allyl-2-alkynylbenzenes
Jeganmohan M and Cheng C-H
Organic Letters, 6(16), 2821-2824 (2004)
Airong Xu et al.
Scientific reports, 9(1), 11518-11518 (2019-08-10)
The utilization of cellulose in industrial applicat is of great significance to sustainable development of human society and reducing dependence on dwindling fossil resources. Nevertheless, this utilization of cellulose has actually been limited due to its insolubilization. Here, novel solvents
Cross-metathesis reactions of allyl chloride with fatty acid methyl esters: Efficient synthesis of a, ?-difunctional chemical intermediates from renewable raw materials.
Jacobs T, et al.
Applied Catalysis A: General, 353(1), 32-35 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service